Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorocinnamoyl chloride, 94%

Chemical Identifiers

Chemical Name or Material 2-Chlorocinnamoyl chloride
CAS 35086-82-9
Molecular Formula C9H6Cl2O
Formula Weight 201.05g/mol
MDL Number MFCD00039311
UN Number UN3261
Beilstein 956921
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1052106

 
alfa aesar™
L10521-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1052114

 
alfa aesar™
L10521-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Melting Point 34° to 39°C
Boiling Point 155° to 160°C (12mmHg)
Quantity 5g
Chemical Name or Material 2-Chlorocinnamoyl chloride
CAS 35086-82-9
Assay 94%
Molecular Formula C9H6Cl2O
Formula Weight 201.05g/mol
MDL Number MFCD00039311
UN Number UN3261
Beilstein 956921
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.