missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorocinnamic acid, predominantly trans, 99%

$59.05 - $154.22

Chemical Identifiers

CAS 3752-25-8
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD00004372
InChI Key KJRRTHHNKJBVBO-AATRIKPKSA-N
Synonym 2-chlorocinnamic acid, o-chlorocinnamic acid, 3-2-chlorophenyl acrylic acid, 2e-3-2-chlorophenyl acrylic acid, e-3-2-chlorophenyl acrylic acid, cinnamic acid, o-chloro, 2e-3-2-chlorophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-chlorophenyl, e-o-chlorocinnamic acid, 3-2-chlorophenyl-2-propenoic acid
PubChem CID 700642
IUPAC Name (E)-3-(2-chlorophenyl)prop-2-enoic acid
SMILES C1=CC=C(C(=C1)C=CC(=O)O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0160614 View Documents Alfa Aesar™
L0160614
25g Each for $59.05
 
AAL0160622 View Documents Alfa Aesar™
L0160622
100g Each for $154.22
 
Specifications

Chemical Identifiers

3752-25-8
182.603
KJRRTHHNKJBVBO-AATRIKPKSA-N
700642
C1=CC=C(C(=C1)C=CC(=O)O)Cl
C9H7ClO2
MFCD00004372
2-chlorocinnamic acid, o-chlorocinnamic acid, 3-2-chlorophenyl acrylic acid, 2e-3-2-chlorophenyl acrylic acid, e-3-2-chlorophenyl acrylic acid, cinnamic acid, o-chloro, 2e-3-2-chlorophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-chlorophenyl, e-o-chlorocinnamic acid, 3-2-chlorophenyl-2-propenoic acid
(E)-3-(2-chlorophenyl)prop-2-enoic acid

Specifications

209°C to 212°C
3752-25-8
MFCD00004372
2-chlorocinnamic acid, o-chlorocinnamic acid, 3-2-chlorophenyl acrylic acid, 2e-3-2-chlorophenyl acrylic acid, e-3-2-chlorophenyl acrylic acid, cinnamic acid, o-chloro, 2e-3-2-chlorophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-chlorophenyl, e-o-chlorocinnamic acid, 3-2-chlorophenyl-2-propenoic acid
C1=CC=C(C(=C1)C=CC(=O)O)Cl
182.603
182.61
2-Chlorocinnamic acid, predominantly trans
25g
C9H7ClO2
1365198
KJRRTHHNKJBVBO-AATRIKPKSA-N
(E)-3-(2-chlorophenyl)prop-2-enoic acid
700642
99%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : GD8450000