Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorobutyryl chloride, tech. 85%


Quantity 5g

Manufacturer: alfa aesar™  L1120706

 View more versions of this product

Catalog No. AAL1120706

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 141
CAS 7623-11-2
Chemical Name or Material 2-Chlorobutyryl chloride
Assay 85%
MDL Number MFCD00041409
UN Number UN3265
Beilstein 741947
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 138° to 140°C
Quantity 5g
Purity Grade Technical Grade
Molecular Formula C4H6Cl2O
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material 2-Chlorobutyryl chloride
CAS 7623-11-2
Purity Grade Technical Grade
Molecular Formula C4H6Cl2O
Formula Weight 141
MDL Number MFCD00041409
UN Number UN3265
Beilstein 741947

Applications
2-Chlorobutyryl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture.

Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.