missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorobutyryl chloride, tech. 85%

Manufacturer:  Alfa Aesar™ L1120714

 View more versions of this product

Catalog No. AAL1120714



Description

Description

Applications
2-Chlorobutyryl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture.
Specifications

Specifications

2-Chlorobutyryl chloride
1.451
138°C to 140°C
7623-11-2
MFCD00041409
741947
Reacts with water.
CC(C(=O)Cl)Cl
126.96
141
Technical
1.236
Moisture sensitive
25g
C3H4Cl2O
UN3265
.alpha.-chloropropionyl chloride, 1,2-dichloro-1-propanone, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 2-chloropropanyl chloride, 2-chloropropionic acid chloride, 2-chloropropionyl chloride, 2-chloropropionylchloride, alpha-chloropropionyl chloride, propanoyl chloride, 2-chloro
JEQDSBVHLKBEIZ-UHFFFAOYNA-N
2-chloropropanoyl chloride
111019
85%

Chemical Identifiers

7623-11-2
126.96
JEQDSBVHLKBEIZ-UHFFFAOYNA-N
111019
CC(C(=O)Cl)Cl
C3H4Cl2O
MFCD00041409
.alpha.-chloropropionyl chloride, 1,2-dichloro-1-propanone, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 2-chloropropanyl chloride, 2-chloropropionic acid chloride, 2-chloropropionyl chloride, 2-chloropropionylchloride, alpha-chloropropionyl chloride, propanoyl chloride, 2-chloro
2-chloropropanoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

Documents