Learn More
2-Chlorobenzyl mercaptan, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 373130050
Specifications
2-Chlorobenzyl mercaptan | |
97.5 | |
1.2200g/mL | |
69°C | |
97.5% min. (GC) | |
C7H7ClS | |
ClC6H4CH2SH | |
5g | |
2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw | |
C1=CC=C(C(=C1)CS)Cl | |
158.65 | |
158.65 |
39718-00-8 | |
100.0 | |
120°C to 121°C (25.0mmHg) | |
Authentic | |
Glass bottle | |
1.5890 to 1.591 | |
MFCD00004868 | |
1.22 | |
WWFIIZLHSNBNTC-UHFFFAOYSA-N | |
(2-chlorophenyl)methanethiol | |
580759 | |
98% |
Chemical Identifiers
39718-00-8 | |
158.65 | |
WWFIIZLHSNBNTC-UHFFFAOYSA-N | |
580759 | |
C1=CC=C(C(=C1)CS)Cl |
C7H7ClS | |
MFCD00004868 | |
2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw | |
(2-chlorophenyl)methanethiol |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 254-602-5
RUO â Research Use Only