Learn More
2-Chlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1582614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chlorobenzyl mercaptan | |
1.22 | |
25 g | |
C7H7ClS | |
UN2810 | |
2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw | |
C1=CC=C(C(=C1)CS)Cl | |
158.643 | |
158.65 |
39718-00-8 | |
120°C to 122°C (25 mmHg) | |
1.591 | |
MFCD00004868 | |
2042183 | |
WWFIIZLHSNBNTC-UHFFFAOYSA-N | |
(2-chlorophenyl)methanethiol | |
580759 | |
97% |
Chemical Identifiers
39718-00-8 | |
158.643 | |
WWFIIZLHSNBNTC-UHFFFAOYSA-N | |
580759 | |
C1=CC=C(C(=C1)CS)Cl |
C7H7ClS | |
MFCD00004868 | |
2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw | |
(2-chlorophenyl)methanethiol |
Safety and Handling
GHS H Statement
H331-H227-H302-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
H227-H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 254-602-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only