Learn More
2-Chlorobenzyl isothiocyanate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1054603
Description
2-Chlorobenzyl isothiocyanate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorobenzyl isothiocyanate is used as pharmaceutical intermediate.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
Specifications
2-Chlorobenzyl isothiocyanate | |
1.149 | |
1 g | |
1.614 | |
MFCD00041110 | |
2936688 | |
It hydrolyzes in water. | |
C1=CC=C(C(=C1)CN=C=S)Cl | |
183.653 | |
183.66 |
18967-44-7 | |
137°C to 138°C (6 mmHg) | |
Irritating | |
C8H6ClNS | |
UN2922 | |
2-chlorobenzyl isothiocyanate, 1-chloro-2-isothiocyanatomethyl benzene, benzene, 1-chloro-2-isothiocyanatomethyl, isothiocyanic acid, o-chlorobenzyl ester, acmc-20any0, wln: scn1r bg, 2-chlorobenzylisothiocyanate, flc 2-chlorobenzyl isothiocyanate, 2-chlorophenyl methanisothiocyanate, 1-chloranyl-2-isothiocyanatomethyl benzene | |
RMVDNJDSLXQPAV-UHFFFAOYSA-N | |
1-chloro-2-(isothiocyanatomethyl)benzene | |
29351 | |
97% |
Chemical Identifiers
18967-44-7 | |
183.653 | |
RMVDNJDSLXQPAV-UHFFFAOYSA-N | |
29351 | |
C1=CC=C(C(=C1)CN=C=S)Cl |
C8H6ClNS | |
MFCD00041110 | |
2-chlorobenzyl isothiocyanate, 1-chloro-2-isothiocyanatomethyl benzene, benzene, 1-chloro-2-isothiocyanatomethyl, isothiocyanic acid, o-chlorobenzyl ester, acmc-20any0, wln: scn1r bg, 2-chlorobenzylisothiocyanate, flc 2-chlorobenzyl isothiocyanate, 2-chlorophenyl methanisothiocyanate, 1-chloranyl-2-isothiocyanatomethyl benzene | |
1-chloro-2-(isothiocyanatomethyl)benzene |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H331-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 000-000-0
RTECSNumber : NX8460000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only