Please login to your online account to display your discounted pricing

2-chlorobenzyl bromide, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC278931000

 View more versions of this product

Catalog No. AC278931000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 205.49
Physical Form Liquid
Quantity 100g
Color Clear yellow
Density 1.586g/mL
Beilstein 05, 155
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.5910 to 1.5930 (20°C, 589nm)
Chemical Name or Material 2-chlorobenzyl bromide
Molecular Formula C7H7BrCl
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Assay Percent Range 97%
MDL Number MFCD00000566
CAS 611-17-6
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula ClC6H4CH2Br

Specifications

Chemical Name or Material 2-chlorobenzyl bromide
CAS 611-17-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 97%
Molecular Formula C7H7BrCl
Linear Formula ClC6H4CH2Br
Formula Weight 205.49
MDL Number MFCD00000566
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents