Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorobenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 2-Chlorobenzyl bromide
CAS 611-17-6
Molecular Formula C7H6BrCl
Formula Weight 205.49
MDL Number MFCD00000566
UN Number UN3265
Beilstein 386264
Synonym alpha-Bromo-2-chlorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1212906

 
alfa aesar™
A1212906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1212914

 
alfa aesar™
A1212914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1212922

 
alfa aesar™
A1212922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.49
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material 2-Chlorobenzyl bromide
Odor Pungent
Assay 97%
Molecular Formula C7H6BrCl
MDL Number MFCD00000566
CAS 611-17-6
UN Number UN3265
Beilstein 386264
Synonym alpha-Bromo-2-chlorotoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point 108°C (226°F)
Boiling Point 103° to 104°C (10mmHg)
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.