missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorobenzyl bromide, 97%

Manufacturer:  Alfa Aesar™ A1212922

 View more versions of this product

Catalog No. AAA1212922


 

Specifications

Specifications

2-Chlorobenzyl bromide
103°C to 104°C (10mmHg)
611-17-6
97%
MFCD00000566
386264
PURSZYWBIQIANP-UHFFFAOYSA-N
1-(bromomethyl)-2-chlorobenzene
11905
108°C (226°F)
100g
Pungent
C7H6BrCl
UN3265
alpha-Bromo-2-chlorotoluene
C1=CC=C(C(=C1)CBr)Cl
205.479
205.49

Chemical Identifiers

611-17-6
205.479
PURSZYWBIQIANP-UHFFFAOYSA-N
11905
C7H6BrCl
MFCD00000566
alpha-Bromo-2-chlorotoluene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents