missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chlorobenzyl alcohol, 99%, ACROS Organics™
Chemical Identifiers
CAS | 17849-38-6 |
---|---|
Molecular Formula | C7H7ClO |
Molecular Weight (g/mol) | 142.58 |
MDL Number | MFCD00004604 |
InChI Key | MBYQPPXEXWRMQC-UHFFFAOYSA-N |
Synonym | 2-chloro-phenyl-methanol, 2-chlorobenzenemethanol, 2-chlorobenzyl alcohol, 2-chlorobenzylalcohol, 2-chlorophenyl methan-1-ol, 2-chlorophenyl methanol, benzenemethanol, 2-chloro, benzenemethanol, ar-chloro, benzyl alcohol, o-chloro, o-chlorobenzyl alcohol |
PubChem CID | 28810 |
IUPAC Name | (2-chlorophenyl)methanol |
SMILES | C1=CC=C(C(=C1)CO)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC163420100
|
Thermo Fisher Scientific
163420100 |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC163420500
|
Thermo Fisher Scientific
163420500 |
50g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
17849-38-6 | |
142.58 | |
MBYQPPXEXWRMQC-UHFFFAOYSA-N | |
28810 | |
C1=CC=C(C(=C1)CO)Cl |
C7H7ClO | |
MFCD00004604 | |
2-chloro-phenyl-methanol, 2-chlorobenzenemethanol, 2-chlorobenzyl alcohol, 2-chlorobenzylalcohol, 2-chlorophenyl methan-1-ol, 2-chlorophenyl methanol, benzenemethanol, 2-chloro, benzenemethanol, ar-chloro, benzyl alcohol, o-chloro, o-chlorobenzyl alcohol | |
(2-chlorophenyl)methanol |
Specifications
Authentic | |
227°C | |
69°C to 72°C | |
17849-38-6 | |
100.0 | |
C7H7ClO | |
MFCD00004604 | |
2-chloro-phenyl-methanol, 2-chlorobenzenemethanol, 2-chlorobenzyl alcohol, 2-chlorobenzylalcohol, 2-chlorophenyl methan-1-ol, 2-chlorophenyl methanol, benzenemethanol, 2-chloro, benzenemethanol, ar-chloro, benzyl alcohol, o-chloro, o-chlorobenzyl alcohol | |
MBYQPPXEXWRMQC-UHFFFAOYSA-N | |
(2-chlorophenyl)methanol | |
28810 | |
Fine Crystalline Powder | |
2-Chlorobenzyl alcohol, 99% |
Glass bottle | |
White | |
10g | |
98.5 | |
98.5% min. (GC) | |
ClC6H4CH2OH | |
06, 444 | |
Solubility in water: limited soluble in water. Other solubilities: 1g/10mL in methanol | |
C1=CC=C(C(=C1)CO)Cl | |
142.58 | |
142.58 | |
99% |
Safety and Handling
EINECSNumber : 241-801-7
TSCA : TSCA