Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorobenzyl alcohol, 98+%

$66.91 - $879.95

Chemical Identifiers

Chemical Name or Material 2-Chlorobenzyl alcohol
CAS 17849-38-6
Molecular Formula C7H7ClO
Formula Weight 142.59
MDL Number MFCD00004604
Beilstein 2042182
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1365714

  SDS
alfa aesar™
A1365714
25g Each for $66.91

AAA1365722

  SDS
alfa aesar™
A1365722
100g Each for $232.04

AAA1365736

  SDS
alfa aesar™
A1365736
500g Each for $879.95
Description & Specifications

Specifications

Formula Weight 142.59
Chemical Name or Material 2-Chlorobenzyl alcohol
Beilstein 2042182
Boiling Point 227°C
Melting Point 69° to 71°C
CAS 17849-38-6
Assay ≥98%
Molecular Formula C7H7ClO
Quantity 25g
MDL Number MFCD00004604