Learn More
2-Chlorobenzoyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1478522
Description
2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.
Solubility
Soluble in acetone, ether, and alcohol. Solubility in water it decomposes.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong bases, alcohols, oxidizing agents.
Specifications
2-Chlorobenzoyl chloride | |
-4°C | |
237°C to 239°C | |
Acrid | |
1.5718 | |
100 g | |
386435 | |
benzoyl chloride, 2-chloro, o-chlorobenzoyl chloride, chlorobenzoyl chloride, benzoyl chloride, o-chloro, ococ, 2-chloro-benzoyl chloride, unii-jr29a4n74x, o-chlorobenzoylchloride, 2-chlorobenzoylchloride, 2-chlorobenzoic acid chloride | |
ONIKNECPXCLUHT-UHFFFAOYSA-N | |
2-chlorobenzoyl chloride | |
69110 | |
175.01 |
609-65-4 | |
1.379 | |
125°C (257°F) | |
C7H4Cl2O | |
MFCD00000660 | |
UN3265 | |
Moisture sensitive | |
Soluble in acetone,ether,and alcohol. Solubility in water it decomposes. | |
ClC(=O)C1=CC=CC=C1Cl | |
175.01 | |
CHEBI:60719 | |
97% |
Chemical Identifiers
609-65-4 | |
175.01 | |
ONIKNECPXCLUHT-UHFFFAOYSA-N | |
69110 | |
2-chlorobenzoyl chloride |
C7H4Cl2O | |
MFCD00000660 | |
benzoyl chloride, 2-chloro, o-chlorobenzoyl chloride, chlorobenzoyl chloride, benzoyl chloride, o-chloro, ococ, 2-chloro-benzoyl chloride, unii-jr29a4n74x, o-chlorobenzoylchloride, 2-chlorobenzoylchloride, 2-chlorobenzoic acid chloride | |
CHEBI:60719 | |
ClC(=O)C1=CC=CC=C1Cl |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 210-194-0
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only