Learn More
2-Chlorobenzoxazole, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 404530500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Chlorobenzoxazole | |
615-18-9 | |
100.0 | |
200.0°C | |
Authentic | |
Glass bottle | |
1.5655 to 1.5675 | |
50g | |
2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole | |
ClC1=NC2=CC=CC=C2O1 | |
153.57 | |
153.57 |
99% | |
98.5 | |
1.3500g/mL | |
85°C | |
97.5% min. (GC) | |
C7H4ClNO | |
MFCD00005766 | |
1.35 | |
BBVQDWDBTWSGHQ-UHFFFAOYSA-N | |
2-chloro-1,3-benzoxazole | |
11986 | |
98% |
Chemical Identifiers
615-18-9 | |
153.57 | |
BBVQDWDBTWSGHQ-UHFFFAOYSA-N | |
11986 | |
ClC1=NC2=CC=CC=C2O1 |
C7H4ClNO | |
MFCD00005766 | |
2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole | |
2-chloro-1,3-benzoxazole |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
EINECSNumber : 210-414-5
RTECSNumber : DM4680000
TSCA : TSCA
RUO â Research Use Only