Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorobenzotrichloride, 99%

Chemical Identifiers

Chemical Name or Material 2-Chlorobenzotrichloride
CAS 2136-89-2
Molecular Formula C7H4Cl4
Formula Weight 229.92
MDL Number MFCD00000788
UN Number UN3261
Synonym alpha,alpha,alpha,2-Tetrachlorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2442814

  SDS
alfa aesar™
B2442814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2442822

  SDS
alfa aesar™
B2442822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.92
TSCA TSCA
Health Hazard 2 GHS H Statement
H301-H314-H318-H351
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
Melting Point 28° to 31°C
MDL Number MFCD00000788
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 260° to 264°C
Molecular Formula C7H4Cl4
Synonym alpha,alpha,alpha,2-Tetrachlorotoluene
CAS 2136-89-2
Quantity 25g
UN Number UN3261
Assay 99%
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Chemical Name or Material 2-Chlorobenzotrichloride
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H351
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.