missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorobenzophenone, 97%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 186631000

 View more versions of this product

Catalog No. AC186631000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
216.67
White to yellow
07, 419
96.0
MFCD00000558
2-Chlorobenzophenone
ClC6H4COC6H5
Authentic
1% max.
Crystalline Powder
100g
42° to 47°C
1191489
100.0
C13H9ClO
97%
97%
0.2% max.

Chemical Identifiers

1191489
100.0
C13H9ClO
216.67
2-Chlorobenzophenone, 97%
96.0
97%
ClC6H4COC6H5
MFCD00000558
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 225-936-9

RTECSNumber : PC4945633

TSCA : TSCA