missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorobenzophenone, 97%, Acros Organics

Chemical Identifiers

CAS 1191489
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C13H9ClO
Linear Formula ClC6H4COC6H5
Formula Weight 216.67
MDL Number MFCD00000558
Chemical Name or Material 2-Chlorobenzophenone, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC186631000 SDS Thermo Fisher Scientific
186631000
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1191489
100.0
C13H9ClO
216.67
2-Chlorobenzophenone, 97%
96.0
97%
ClC6H4COC6H5
MFCD00000558
≥96.0%

Specifications

Crystalline Powder
100g
42° to 47°C
1191489
100.0
C13H9ClO
ClC6H4COC6H5
0.2% max.
2-Chlorobenzophenone, 97%
216.67
White to yellow
07, 419
96.0
MFCD00000558
97%
Authentic
1% max.
≥96.0%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 225-936-9

RTECSNumber : PC4945633

TSCA : TSCA