Please login to your online account to display your discounted pricing

2-Chlorobenzoic hydrazide, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chlorobenzoic hydrazide
Name Note 97%
CAS 5814-05-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H7ClN2O
Linear Formula ClC6H4CONHNH2
Formula Weight 170.6
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369220250

  SDS
thermo fisher scientific
AC369220250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 170.6
Physical Form Crystalline Powder
Quantity 25g
Color White to slightly beige
Melting Point 113° to 118°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
MDL Number MFCD00007597
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula ClC6H4CONHNH2
Chemical Name or Material 2-Chlorobenzoic hydrazide
CAS 5814-05-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Molecular Formula C7H7ClN2O
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.