Learn More
2-Chlorobenzoic acid, 98+%
CAS: 118-91-2 | C7H5ClO2 | 156.565 g/mol
Supplier: Thermo Scientific Chemicals A108320B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Chlorobenzoic acid | |
| 138°C to 142°C | |
| 284°C to 286°C | |
| C7H5ClO2 | |
| 1000 g | |
| 14,2124 | |
| IKCLCGXPQILATA-UHFFFAOYSA-N | |
| 2-chlorobenzoic acid | |
| 8374 | |
| 156.57 |
| 118-91-2 | |
| 1.544 | |
| 173°C (343°F) | |
| MFCD00002412 | |
| 907340 | |
| o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate | |
| C1=CC=C(C(=C1)C(=O)O)Cl | |
| 156.565 | |
| CHEBI:30793 | |
| ≥98% |
Chemical Identifiers
| 118-91-2 | |
| 156.565 | |
| IKCLCGXPQILATA-UHFFFAOYSA-N | |
| 8374 | |
| 2-chlorobenzoic acid |
| C7H5ClO2 | |
| MFCD00002412 | |
| o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate | |
| CHEBI:30793 | |
| C1=CC=C(C(=C1)C(=O)O)Cl |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H500
EINECSNumber : 204-285-4
RTECSNumber : DG4976000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only