Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorobenzhydrazide, 98+%

$38.49 - $111.91

Chemical Identifiers

Chemical Name or Material 2-Chlorobenzhydrazide
CAS 5814-05-1
Molecular Formula C7H7ClN2O
Formula Weight 170.6
MDL Number MFCD00007597
Beilstein 972229
Synonym 2-Chlorobenzoic acid hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1136106

 
alfa aesar™
A1136106
5g Each for $38.49

AAA1136114

 
alfa aesar™
A1136114
25g Each for $111.91
Description & Specifications

Specifications

Formula Weight 170.6
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Chlorobenzhydrazide
Assay ≥98%
Molecular Formula C7H7ClN2O
MDL Number MFCD00007597
CAS 5814-05-1
Beilstein 972229
Synonym 2-Chlorobenzoic acid hydrazide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 116° to 120°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.