missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorobenzenesulfonyl chloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC357460250

 View more versions of this product

Catalog No. AC357460250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Chlorobenzenesulfonyl chloride
Clear yellow to orange-red
2905-23-9
100.0
Glass bottle
Liquid or Low Melting Solid
98%
ClC6H4SO2Cl
MFCD00051520
1.5480g/mL
25g
97.5
Authentic
1.5745 to 1.5765 (20°C, 589nm)
≥97.5%
C6H4Cl2O2S
211.07

Chemical Identifiers

2905-23-9
100.0
C6H4Cl2O2S
211.07
2-Chlorobenzenesulfonyl chloride
97.5
98%
ClC6H4SO2Cl
MFCD00051520
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger