missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorobenzenesulfonamide, 98%

Manufacturer:  Alfa Aesar™ L0908206

 View more versions of this product

Catalog No. AAL0908206



Specifications

Specifications

2-Chlorobenzenesulfonamide
5g
C6H6ClNO2S
2937540
JCCBZCMSYUSCFM-UHFFFAOYSA-N
2-chlorobenzenesulfonamide
81410
98%
189°C to 191°C
6961-82-6
MFCD00051974
o-chlorobenzenesulfonamide, 2-chlorobenzene-1-sulfonamide, benzenesulfonamide, 2-chloro, unii-4bt2i28a0c, benzenesulfonamide, o-chloro, 2-chlorobenzenesulphonamide, 2-chloro-benzenesulfonamide, chlorobenzenesulfonamide, o-chlorobenzenesulphonamide, benzenesulfonamide, chloro
C1=CC=C(C(=C1)S(=O)(=O)N)Cl
191.629
191.64

Chemical Identifiers

6961-82-6
191.629
JCCBZCMSYUSCFM-UHFFFAOYSA-N
81410
C1=CC=C(C(=C1)S(=O)(=O)N)Cl
C6H6ClNO2S
MFCD00051974
o-chlorobenzenesulfonamide, 2-chlorobenzene-1-sulfonamide, benzenesulfonamide, 2-chloro, unii-4bt2i28a0c, benzenesulfonamide, o-chloro, 2-chlorobenzenesulphonamide, 2-chloro-benzenesulfonamide, chlorobenzenesulfonamide, o-chlorobenzenesulphonamide, benzenesulfonamide, chloro
2-chlorobenzenesulfonamide
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents