missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorobenzaldehyde 99%, ACROS Organics™

Manufacturer:  Acros Organics 154615000

 View more versions of this product

Catalog No. AC154615000



Specifications

Specifications

2-Chlorobenzaldehyde
Glass bottle
Colorless to Yellow
99%
98.5
99%
ClC6H4CHO
07, 233
FPYUJUBAXZAQNL-UHFFFAOYSA-N
2-chlorobenzaldehyde
6996
Liquid
Authentic
1.5650 to 1.5670 (20°C, 589nm)
500mL
89-98-5
100.0
C7H5ClO
MFCD00003304
o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde
C1=CC=C(C(=C1)C=O)Cl
140.566
140.57
≥98.5%

Chemical Identifiers

89-98-5
140.566
FPYUJUBAXZAQNL-UHFFFAOYSA-N
6996
C1=CC=C(C(=C1)C=O)Cl
C7H5ClO
MFCD00003304
o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde
2-chlorobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 201-956-3

RTECSNumber : CU5075000

TSCA : TSCA

SDS Documents