Please login to your online account to display your discounted pricing

2-Chloroanisole, 98%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  171422500

 View more versions of this product

Catalog No. AC171422500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 142.58
Quantity 250g
Color Clear light yellow
Beilstein 06, 184
EINECS Number 212-167-9
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 2-Chloroanisole
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H7ClO
MDL Number MFCD00000557
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5435 to 1.5455 (20°C, 589nm)
Linear Formula ClC6H4OCH3
CAS 766-51-8
Synonym 1-Chloro-2-methoxybenzene
CAS Min % 97.5

Specifications

Chemical Name or Material 2-Chloroanisole
Name Note 98%
CAS 766-51-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7ClO
Linear Formula ClC6H4OCH3
Formula Weight 142.58
MDL Number MFCD00000557
Synonym 1-Chloro-2-methoxybenzene