Please login to your online account to display your discounted pricing

2-Chloroanisole, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloroanisole
Name Note 98%
CAS 766-51-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7ClO
Linear Formula ClC6H4OCH3
Formula Weight 142.58
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171422500

  SDS
thermo fisher scientific
171422500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 142.58
Physical Form Liquid
Quantity 250g
Color Clear light yellow
Beilstein 06, 184
TSCA TSCA
EINECS Number 212-167-9
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Chemical Name or Material 2-Chloroanisole
Name Note 98%
MDL Number MFCD00000557
Molecular Formula C7H7ClO
CAS 766-51-8
CAS Min % 97.5
Linear Formula ClC6H4OCH3
Refractive Index 1.5435 to 1.5455 (20°C, 589nm)
Synonym 1-Chloro-2-methoxybenzene
Packaging Glass bottle