missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroaniline hydrochloride, 97%

Manufacturer:  Alfa Aesar™ L1348314

 View more versions of this product

Catalog No. AAL1348314



Description

Description

Applications
2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

2-Chloroaniline hydrochloride
~220°C (sublimation)
137-04-2
MFCD00053282
3624280
DRGIDRZFKRLQTE-UHFFFAOYSA-N
2-chloroaniline;hydrochloride
67298
97%
Characteristic
25g
C6H7Cl2N
UN2018
2-chloroaniline hydrochloride, 2-chloroanilinium chloride, benzenamine, 2-chloro-, hydrochloride, benzenamine, 2-chloro-, hydrochloride 1:1, 2-chlorophenylamine, chloride, acmc-209cai, o-chloroaniline hydrochloride, 95-51-2 parent, 2-chloranylaniline hydrochloride, benzenamine,2-chloro-,hydrochloride 1:1
C1=CC=C(C(=C1)N)Cl.Cl
164.029
164.03

Chemical Identifiers

137-04-2
164.029
DRGIDRZFKRLQTE-UHFFFAOYSA-N
67298
C1=CC=C(C(=C1)N)Cl.Cl
C6H7Cl2N
MFCD00053282
2-chloroaniline hydrochloride, 2-chloroanilinium chloride, benzenamine, 2-chloro-, hydrochloride, benzenamine, 2-chloro-, hydrochloride 1:1, 2-chlorophenylamine, chloride, acmc-209cai, o-chloroaniline hydrochloride, 95-51-2 parent, 2-chloranylaniline hydrochloride, benzenamine,2-chloro-,hydrochloride 1:1
2-chloroaniline;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P280h-P273-P309-P310-P302+P352-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID

TSCA : TSCA

Documents