Please login to your online account to display your discounted pricing

2-chloroaniline, 98+%, ACROS Organics™


Quantity 5L
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC108570050

 View more versions of this product

Catalog No. AC108570050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
May cause damage to organs through prolonged or repeated exposure. Toxic if inhaled. Very toxic to aquatic life with long lasting effects. Toxic if swallowed. Toxic in contact with skin.
Health Hazard 3 GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.
Assay ≥98.0%
Physical Form Liquid
Formula Weight 127.57
Quantity 5L
Color Clear pale yellow to brown
Packaging Plastic Jerrycan
Density 1.2100g/mL
Beilstein 12, 597
Merck Index 15, 2118
CAS Min % 98.0
CAS Max % 100.0
Refractive Index 1.5870 to 1.5890 (20°C, 589nm)
Linear Formula ClC6H4NH2
Storage Information The product turns dark during prolonged storage
Infrared Spectrum Authentic
Health Hazard 1 Danger
MDL Number MFCD00007656
CAS 95-51-2
Assay Percent Range 98+%
Chemical Name or Material 2-Chloroaniline
Molecular Formula C6H6ClN
Synonym 2-Chlorobenzenamine

Specifications

Chemical Name or Material 2-Chloroaniline
CAS 95-51-2
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C6H6ClN
Linear Formula ClC6H4NH2
Formula Weight 127.57
MDL Number MFCD00007656
Synonym 2-Chlorobenzenamine
Accessories

Danger

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

GHS H Statement
May cause damage to organs through prolonged or repeated exposure. Toxic if inhaled. Very toxic to aquatic life with long lasting effects. Toxic if swallowed. Toxic in contact with skin.

SDS & Safety Documents