Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroadenine, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloroadenine
CAS 1839-18-5
Formula Weight 169.57
MDL Number MFCD00037925
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH60869ME

  SDS
alfa aesar™
H60869ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.57
Melting Point 384° to 389°C
Chemical Name or Material 2-Chloroadenine
Physical Form Solid
Assay 97%
MDL Number MFCD00037925
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Pale yellow
Quantity 500mg
CAS 1839-18-5
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.