Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroadamantane, 98%

$56.50 - $753.38

Chemical Identifiers

Chemical Name or Material 2-Chloroadamantane
CAS 7346-41-0
Molecular Formula C10H15Cl
Formula Weight 170.68
MDL Number MFCD00075643
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2119706

  SDS
alfa aesar™
B2119706
5g Each for $56.50

AAB2119722

  SDS
alfa aesar™
B2119722
100g Each for $753.38
Description & Specifications

Specifications

Formula Weight 170.68
MDL Number MFCD00075643
Molecular Formula C10H15Cl
CAS 7346-41-0
Chemical Name or Material 2-Chloroadamantane
Assay 98%
Quantity 5g