missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Chloroacetophenone 97.0+%, TCI America™
$77.97 - $360.00
Chemical Identifiers
CAS | 2142-68-9 |
---|---|
Molecular Formula | C8H7ClO |
Molecular Weight (g/mol) | 154.593 |
MDL Number | MFCD00000560 |
InChI Key | ZDOYHCIRUPHUHN-UHFFFAOYSA-N |
Synonym | 2'-chloroacetophenone, 1-2-chlorophenyl ethanone, o-chloroacetophenone, 1-2-chlorophenyl ethan-1-one, ethanone, 1-2-chlorophenyl, acetophenone, 2'-chloro, o-chloroacetophonone, 1-2-chloro-phenyl-ethanone, ethanone, 1-chlorophenyl, 1-acetyl-2-chlorobenzene |
PubChem CID | 72864 |
IUPAC Name | 1-(2-chlorophenyl)ethanone |
SMILES | CC(=O)C1=CC=CC=C1Cl |
Chemical Identifiers
2142-68-9 | |
154.593 | |
ZDOYHCIRUPHUHN-UHFFFAOYSA-N | |
72864 | |
CC(=O)C1=CC=CC=C1Cl |
C8H7ClO | |
MFCD00000560 | |
2'-chloroacetophenone, 1-2-chlorophenyl ethanone, o-chloroacetophenone, 1-2-chlorophenyl ethan-1-one, ethanone, 1-2-chlorophenyl, acetophenone, 2'-chloro, o-chloroacetophonone, 1-2-chloro-phenyl-ethanone, ethanone, 1-chlorophenyl, 1-acetyl-2-chlorobenzene | |
1-(2-chlorophenyl)ethanone |
Specifications
2142-68-9 | |
229°C | |
MFCD00000560 | |
3416 | |
ZDOYHCIRUPHUHN-UHFFFAOYSA-N | |
1-(2-chlorophenyl)ethanone | |
72864 | |
≥97.0% (GC) | |
2′-Chloroacetophenone |
Yellow | |
C8H7ClO | |
25 g | |
2'-chloroacetophenone, 1-2-chlorophenyl ethanone, o-chloroacetophenone, 1-2-chlorophenyl ethan-1-one, ethanone, 1-2-chlorophenyl, acetophenone, 2'-chloro, o-chloroacetophonone, 1-2-chloro-phenyl-ethanone, ethanone, 1-chlorophenyl, 1-acetyl-2-chlorobenzene | |
CC(=O)C1=CC=CC=C1Cl | |
154.593 | |
154.59 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-1241&(3)-2933
RTECSNumber : AM6310000
TSCA : Yes