missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Chloroacetophenone, 98%

Manufacturer:  Alfa Aesar™ A1523230

 View more versions of this product

Catalog No. AAA1523230



Specifications

Specifications

2'-Chloroacetophenone
228°C to 229°C
2142-68-9
MFCD00000560
1858916
ZDOYHCIRUPHUHN-UHFFFAOYSA-N
1-(2-chlorophenyl)ethanone
72864
98%
88°C (190°F)
250g
C8H7ClO
UN3416
2'-chloroacetophenone, 1-2-chlorophenyl ethanone, o-chloroacetophenone, 1-2-chlorophenyl ethan-1-one, ethanone, 1-2-chlorophenyl, acetophenone, 2'-chloro, o-chloroacetophonone, 1-2-chloro-phenyl-ethanone, ethanone, 1-chlorophenyl, 1-acetyl-2-chlorobenzene
CC(=O)C1=CC=CC=C1Cl
154.593
154.6

Chemical Identifiers

2142-68-9
154.593
ZDOYHCIRUPHUHN-UHFFFAOYSA-N
72864
CC(=O)C1=CC=CC=C1Cl
C8H7ClO
MFCD00000560
2'-chloroacetophenone, 1-2-chlorophenyl ethanone, o-chloroacetophenone, 1-2-chlorophenyl ethan-1-one, ethanone, 1-2-chlorophenyl, acetophenone, 2'-chloro, o-chloroacetophonone, 1-2-chloro-phenyl-ethanone, ethanone, 1-chlorophenyl, 1-acetyl-2-chlorobenzene
1-(2-chlorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, LIQUID

RTECSNumber : AM6310000

TSCA : TSCA

Documents