Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Chloroacetophenone, 98%

Chemical Identifiers

Chemical Name or Material 2'-Chloroacetophenone
CAS 2142-68-9
Molecular Formula C8H7ClO
Formula Weight 154.6
MDL Number MFCD00000560
UN Number UN3416
Beilstein 1858916
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA152320B

  SDS
alfa aesar™
A152320B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1523230

  SDS
alfa aesar™
A1523230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1523218

  SDS
alfa aesar™
A1523218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.6
UN Number UN3416
CAS 2142-68-9
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H7ClO
MDL Number MFCD00000560
Health Hazard 1 Danger
Chemical Name or Material 2'-Chloroacetophenone
Beilstein 1858916
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, LIQUID
Health Hazard 2 GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Quantity 1000g
Boiling Point 228° to 229°C
Flash Point 88°C (190°F)
Assay 98%
RTECS Number AM6310000
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.