Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroacetophenone, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloroacetophenone
CAS 532-27-4
Molecular Formula C8H7ClO
Formula Weight 154.60g/mol
MDL Number MFCD00000933
UN Number UN1697
Beilstein 507950
Merck Index 14,2115
Synonym alpha-Chloroacetophenone; Phenacyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1074936

 
alfa aesar™
A10749-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500g
Boiling Point 243° to 244°C
Melting Point 52° to 56°C
Quantity 500g
Flash Point 118°C (244°F)
Odor Irritating
Chemical Name or Material 2-Chloroacetophenone
CAS 532-27-4
Assay 98%
Molecular Formula C8H7ClO
Formula Weight 154.60g/mol
MDL Number MFCD00000933
UN Number UN1697
Beilstein 507950
Merck Index 14,2115
Synonym alpha-Chloroacetophenone; Phenacyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H331-H319-H317-H335
Fatal if swallowed.
Toxic if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, SOLID
RTECS Number AM6300000
TSCA TSCA