Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Chloroacetoacetanilide, 98%

$265.19 - $265.19

Chemical Identifiers

Chemical Name or Material 2'-Chloroacetoacetanilide
CAS 93-70-9
Molecular Formula C10H10ClNO2
Formula Weight 211.65
MDL Number MFCD00018224
Synonym N-(2-Chlorophenyl)-3-oxobutyramide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1867422

  SDS
alfa aesar™
A1867422
100g Each for $265.19
Description & Specifications

Specifications

Formula Weight 211.65
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
RTECS Number AK4300000
TSCA TSCA
CAS 93-70-9
Quantity 100g
MDL Number MFCD00018224
Melting Point 104° to 106°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2'-Chloroacetoacetanilide
Synonym N-(2-Chlorophenyl)-3-oxobutyramide
Molecular Formula C10H10ClNO2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.