Please login to your online account to display your discounted pricing

2'-Chloroacetanilide, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  249430050

 View more versions of this product

Catalog No. AC249430050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 169.61
Quantity 5g
Color Slightly beige
EINECS Number 208-555-2
TSCA TSCA
Melting Point 85° to 88°C
CAS Max % 100.0
CAS Min % 99.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Name Note 99+%
Molecular Formula C8H8ClNO
CAS 533-17-5
Chemical Name or Material 2';-Chloroacetanilide
MDL Number MFCD00045169
Assay Percent Range 99+%
Infrared Spectrum Authentic
Synonym N-(2-Chlorophenyl)acetamide

Specifications

Chemical Name or Material 2';-Chloroacetanilide
Name Note 99+%
CAS 533-17-5
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H8ClNO
Formula Weight 169.61
MDL Number MFCD00045169
Synonym N-(2-Chlorophenyl)acetamide
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents