missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloroacetamide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 148415000

 View more versions of this product

Catalog No. AC148415000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloroacetamide
79-07-2
100.0
White
224.5°C
Authentic
Plastic bottle
ClCH2CONH2
500 g
13,70
15, 2110
Solubility in water: 90g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether
NC(=O)CCl
93.51
93.51
Crystalline Powder
98%
97.5
116.0°C to 120.0°C
0.8400g/mL
170°C
97.5% min. (GC)
C2H4ClNO
MFCD00008027
02, 199
0.84
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide
VXIVSQZSERGHQP-UHFFFAOYSA-N
2-chloroacetamide
6580
98%

Chemical Identifiers

79-07-2
93.51
VXIVSQZSERGHQP-UHFFFAOYSA-N
6580
NC(=O)CCl
C2H4ClNO
MFCD00008027
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide
2-chloroacetamide
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
May cause an allergic skin reaction.
Suspected of damaging fertility.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-174-2

RTECSNumber : AB5075000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only