missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloroacetamide 98%, ACROS Organics™

Manufacturer:  Acros Organics 148410010

 View more versions of this product

Catalog No. AC148410010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloroacetamide
170°C
Plastic bottle
0.2% max.
White
1kg
79-07-2
100.0
C2H4ClNO
MFCD00008027
13,70
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide
VXIVSQZSERGHQP-UHFFFAOYSA-N
2-chloroacetamide
6580
Crystalline Powder
0.8400g/mL
Authentic
0.84
224.5°C
116.0°C to 120.0°C
98%
97.5
98%
ClCH2CONH2
02, 199
15, 2110
Solubility in water: 90g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether
C(C(=O)N)Cl
93.51
93.51
98%

Chemical Identifiers

79-07-2
93.51
VXIVSQZSERGHQP-UHFFFAOYSA-N
6580
C(C(=O)N)Cl
C2H4ClNO
MFCD00008027
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide
2-chloroacetamide
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
May cause an allergic skin reaction.
Suspected of damaging fertility.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-174-2

RTECSNumber : AB5075000

TSCA : TSCA

SDS Documents