Learn More
2-Chloroacetamide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1523830
Description
2-Chloroacetamide is used as surface-active agent in home laundry, cleaning products & household formulated cleaners, other industrial uses, petroleum production, polymer processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloroacetamide is used as surface-active agent in home laundry, cleaning products & household formulated cleaners, other industrial uses, petroleum production, polymer processing.
Solubility
Soluble in water(90g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
2-Chloroacetamide | |
116°C to 119°C | |
170°C (338°F) | |
C2H4ClNO | |
250 g | |
878191 | |
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide | |
VXIVSQZSERGHQP-UHFFFAOYSA-N | |
2-chloroacetamide | |
6580 | |
≥98% |
79-07-2 | |
220°C (decomposition) | |
Weak | |
MFCD00008027 | |
UN2811 | |
14,2110 | |
Soluble in water(90g/L). | |
NC(=O)CCl | |
93.51 | |
93.51 |
Chemical Identifiers
79-07-2 | |
93.51 | |
VXIVSQZSERGHQP-UHFFFAOYSA-N | |
6580 | |
NC(=O)CCl |
C2H4ClNO | |
MFCD00008027 | |
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide | |
2-chloroacetamide |
Safety and Handling
GHS H Statement
H301-H361-H317
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P308+P313-P330-P333+P313-P363-P501c
H301-H317-H361f-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 201-174-2
RTECSNumber : AB5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only