missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-p-phenylenediamine sulfate, 95%

$104.73 - $407.12

Chemical Identifiers

CAS 61702-44-1
Molecular Formula C6H9ClN2O4S
Molecular Weight (g/mol) 240.658
MDL Number MFCD00013004
InChI Key GQFGHCRXPLROOF-UHFFFAOYSA-N
Synonym 1,4-benzenediamine, 2-chloro-, sulfate, 2-chloro-1,4-benzenediamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 2-cl-p-pd, fourrine 81, fourrine so, renal so
PubChem CID 22584
ChEBI CHEBI:76597
IUPAC Name 2-chlorobenzene-1,4-diamine;sulfuric acid
SMILES C1=CC(=C(C=C1N)Cl)N.OS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2402930
View Documents
Alfa Aesar™
B2402930
250g Each for $104.73
 
AAB240290B
View Documents
Alfa Aesar™
B240290B
1000g Each for $407.12
 
Specifications

Chemical Identifiers

61702-44-1
240.658
GQFGHCRXPLROOF-UHFFFAOYSA-N
22584
2-chlorobenzene-1,4-diamine;sulfuric acid
C6H9ClN2O4S
MFCD00013004
1,4-benzenediamine, 2-chloro-, sulfate, 2-chloro-1,4-benzenediamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 2-cl-p-pd, fourrine 81, fourrine so, renal so
CHEBI:76597
C1=CC(=C(C=C1N)Cl)N.OS(=O)(=O)O

Specifications

251°C to 253°C
61702-44-1
MFCD00013004
8441373
GQFGHCRXPLROOF-UHFFFAOYSA-N
2-chlorobenzene-1,4-diamine;sulfuric acid
22584
240.67
2-Chloro-p-phenylenediamine sulfate
250g
C6H9ClN2O4S
UN2811
1,4-benzenediamine, 2-chloro-, sulfate, 2-chloro-1,4-benzenediamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 2-cl-p-pd, fourrine 81, fourrine so, renal so
C1=CC(=C(C=C1N)Cl)N.OS(=O)(=O)O
240.658
CHEBI:76597
95%
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 262-915-3

RTECSNumber : CZ1581000

TSCA : No

Recommended Storage : Ambient temperatures