missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-N,N-dimethylacetamide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438210050

 View more versions of this product

Catalog No. AC438210050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-chloro-N, N-dimethylacetamide
>110°C
Glass bottle
98°C to 100°C (11.0 Torr)
15°C to 16°C
2675-89-0
100.0
C4H8ClNO
2-chloro-dimethylacetamide, 2-chloro-n, n-dimethylacetamide, 2-chlorodimethyl acetamide, 2-chlorodimethylacetamide, acetamide, 2-chloro-n,n-dimethyl, acmc-209gsx, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide, n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide
XBPPLECAZBTMMK-UHFFFAOYSA-N
2-chloro-N,N-dimethylacetamide
75886
Liquid
1.1820g/mL
Authentic
1.182
Colorless to Yellow
5mL
96.0
96% min. (GC)
ClCH2CON(CH3)2
Solubility in water: in water:(20°C) soluble.,in ethanol:(20°C) soluble
CN(C)C(=O)CCl
121.57
121.57
97%

Chemical Identifiers

2675-89-0
121.57
2-chloro-dimethylacetamide, 2-chloro-n, n-dimethylacetamide, 2-chlorodimethyl acetamide, 2-chlorodimethylacetamide, acetamide, 2-chloro-n,n-dimethyl, acmc-209gsx, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide, n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide
2-chloro-N,N-dimethylacetamide
C4H8ClNO
XBPPLECAZBTMMK-UHFFFAOYSA-N
75886
CN(C)C(=O)CCl
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 220-224-4

SDS Documents