missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-chloro-N,N-dimethylacetamide, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438210250
Specifications
2-chloro-N, N-dimethylacetamide | |
>110°C | |
Glass bottle | |
98°C to 100°C (11.0 Torr) | |
15°C to 16°C | |
2675-89-0 | |
100.0 | |
C4H8ClNO | |
2-chloro-dimethylacetamide, 2-chloro-n, n-dimethylacetamide, 2-chlorodimethyl acetamide, 2-chlorodimethylacetamide, acetamide, 2-chloro-n,n-dimethyl, acmc-209gsx, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide, n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide | |
XBPPLECAZBTMMK-UHFFFAOYSA-N | |
2-chloro-N,N-dimethylacetamide | |
75886 | |
Liquid |
1.1820g/mL | |
Authentic | |
1.182 | |
Colorless to Yellow | |
25mL | |
96.0 | |
96% min. (GC) | |
ClCH2CON(CH3)2 | |
Solubility in water: in water:(20°C) soluble.,in ethanol:(20°C) soluble | |
CN(C)C(=O)CCl | |
121.57 | |
121.57 | |
97% |
Chemical Identifiers
2675-89-0 | |
121.57 | |
2-chloro-dimethylacetamide, 2-chloro-n, n-dimethylacetamide, 2-chlorodimethyl acetamide, 2-chlorodimethylacetamide, acetamide, 2-chloro-n,n-dimethyl, acmc-209gsx, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide, n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide | |
2-chloro-N,N-dimethylacetamide |
C4H8ClNO | |
XBPPLECAZBTMMK-UHFFFAOYSA-N | |
75886 | |
CN(C)C(=O)CCl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 220-224-4