missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-N,N-dimethylacetamide, 97%, Thermo Scientific Chemicals

$137.97 - $448.22

Chemical Identifiers

CAS 2675-89-0
Molecular Formula C4H8ClNO
MDL Number MFCD00039360
InChI Key XBPPLECAZBTMMK-UHFFFAOYSA-N
PubChem CID 75886
IUPAC Name 2-chloro-N,N-dimethylacetamide
SMILES CN(C)C(=O)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50229MD
View Documents Promotion Details
Thermo Scientific Chemicals
H50229MD
250 mg
Each for $137.97
 
AAH5022903
View Documents Promotion Details
Thermo Scientific Chemicals
H5022903
1 g
Each for $448.22
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2675-89-0
MFCD00039360
75886
CN(C)C(=O)CCl
C4H8ClNO
XBPPLECAZBTMMK-UHFFFAOYSA-N
2-chloro-N,N-dimethylacetamide

Specifications

2675-89-0
98°C to 100°C (11 mmHg)
1.479
MFCD00039360
969568
XBPPLECAZBTMMK-UHFFFAOYSA-N
2-chloro-N,N-dimethylacetamide
75886
97%
1.182
C4H8ClNO
ClCH2CON(CH3)2
250 mg
n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide, acetamide, 2-chloro-n,n-dimethyl, 2-chloro-n, n-dimethylacetamide, acmc-209gsx, 2-chlorodimethylacetamide, 2-chloro-dimethylacetamide, 2-chlorodimethyl acetamide, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide
CN(C)C(=O)CCl
121.564
121.57
2-Chloro-N,N-dimethylacetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 220-224-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only