Alfa Aesar™ 2-Chloro-N,N-dimethyl-6-nitroaniline, 96%
Manufacturer:  Alfa Aesar™ H63288MD
 View more versions of this product
Catalog No. AAH63288MD
Specifications
Specifications
2-Chloro-N,N-dimethyl-6-nitroaniline
96994-75-1
UN2811
2-chloro-6-nitro-n,n-dimethylaniline, 2-chloro-n,n-dimethyl-6-nitrobenzenamine, 2-chloro-6-nitro-phenyl-dimethyl-amine
CN(C)C1=C(C=CC=C1Cl)[N+](=O)[O-]
200.622
200.62
96%
250mg
C8H9ClN2O2
3269421
AMHCHWZVYGFSNK-UHFFFAOYSA-N
2-chloro-N,N-dimethyl-6-nitroaniline
13418267
Solid
Chemical Identifiers
96994-75-1
200.622
2-chloro-6-nitro-n,n-dimethylaniline, 2-chloro-n,n-dimethyl-6-nitrobenzenamine, 2-chloro-6-nitro-phenyl-dimethyl-amine
2-chloro-N,N-dimethyl-6-nitroaniline
C8H9ClN2O2
AMHCHWZVYGFSNK-UHFFFAOYSA-N
13418267
CN(C)C1=C(C=CC=C1Cl)[N+](=O)[O-]
Safety and Handling
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
SDS Documents