Learn More
2-Chloro-N,N-dimethyl-4-nitroaniline, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H63488MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Chloro-N,N-dimethyl-4-nitroaniline | |
C8H9ClN2O2 | |
250 mg | |
2-chloro-nn-dimethyl-4-nitroaniline, benzenamine, 2-chloro-n,n-dimethyl-4-nitro, n,n-dimethyl-4-nitro-2-chloroaniline, 2-chloro-4-nitrophenyl dimethylamine, 3-chloro-4-dimethylamino nitrobenzene, 3-chloro-4-dimethylaminonitrobenzene, 2-chloro-n,n-dimethyl-4-nitrobenzenamine, benzenamine,2-chloro-n,n-dimethyl-4-nitro, 2-chloro-4-nitro-n,n-dimethylaniline | |
CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])Cl | |
200.622 | |
200.62 | |
Solid |
6213-19-0 | |
MFCD00043604 | |
UN2811 | |
OZKAWTHGBGLZKC-UHFFFAOYSA-N | |
2-chloro-N,N-dimethyl-4-nitroaniline | |
80343 | |
96% |
Chemical Identifiers
6213-19-0 | |
200.622 | |
OZKAWTHGBGLZKC-UHFFFAOYSA-N | |
80343 | |
CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])Cl |
C8H9ClN2O2 | |
MFCD00043604 | |
2-chloro-nn-dimethyl-4-nitroaniline, benzenamine, 2-chloro-n,n-dimethyl-4-nitro, n,n-dimethyl-4-nitro-2-chloroaniline, 2-chloro-4-nitrophenyl dimethylamine, 3-chloro-4-dimethylamino nitrobenzene, 3-chloro-4-dimethylaminonitrobenzene, 2-chloro-n,n-dimethyl-4-nitrobenzenamine, benzenamine,2-chloro-n,n-dimethyl-4-nitro, 2-chloro-4-nitro-n,n-dimethylaniline | |
2-chloro-N,N-dimethyl-4-nitroaniline |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 228-280-1
TSCA : Yes
Recommended Storage : Ambient temperatures