missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-N,N-dimethyl-4-nitroaniline, 96%

Manufacturer:  Alfa Aesar™ H6348806

 View more versions of this product

Catalog No. AAH6348806



Specifications

Specifications

5g
C8H9ClN2O2
UN2811
OZKAWTHGBGLZKC-UHFFFAOYSA-N
2-chloro-N,N-dimethyl-4-nitroaniline
80343
Solid
6213-19-0
MFCD00043604
2-chloro-4-nitro-n,n-dimethylaniline, 2-chloro-4-nitrophenyl dimethylamine, 2-chloro-n,n-dimethyl-4-nitrobenzenamine, 2-chloro-nn-dimethyl-4-nitroaniline, 3-chloro-4-dimethylamino nitrobenzene, 3-chloro-4-dimethylaminonitrobenzene, benzenamine, 2-chloro-n,n-dimethyl-4-nitro, benzenamine,2-chloro-n,n-dimethyl-4-nitro, n,n-dimethyl-4-nitro-2-chloroaniline
CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])Cl
200.622
200.62
96%

Chemical Identifiers

6213-19-0
200.622
OZKAWTHGBGLZKC-UHFFFAOYSA-N
80343
CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])Cl
C8H9ClN2O2
MFCD00043604
2-chloro-4-nitro-n,n-dimethylaniline, 2-chloro-4-nitrophenyl dimethylamine, 2-chloro-n,n-dimethyl-4-nitrobenzenamine, 2-chloro-nn-dimethyl-4-nitroaniline, 3-chloro-4-dimethylamino nitrobenzene, 3-chloro-4-dimethylaminonitrobenzene, benzenamine, 2-chloro-n,n-dimethyl-4-nitro, benzenamine,2-chloro-n,n-dimethyl-4-nitro, n,n-dimethyl-4-nitro-2-chloroaniline
2-chloro-N,N-dimethyl-4-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 228-280-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents