Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-N,N-diethyl-6-nitroaniline, 96%

Chemical Identifiers

Chemical Name or Material 2-Chloro-N,N-diethyl-6-nitroaniline
CAS 133387-28-7
Formula Weight 228.68
UN Number UN2811
Beilstein 3312621
Synonym (2-Chloro-6-nitrophenyl)diethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6375403

  SDS
alfa aesar™
H6375403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6375406

  SDS
alfa aesar™
H6375406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63754MD

  SDS
alfa aesar™
H63754MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 228.68
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Chloro-N,N-diethyl-6-nitroaniline
Physical Form Solid
Assay 96%
CAS 133387-28-7
UN Number UN2811
Beilstein 3312621
Synonym (2-Chloro-6-nitrophenyl)diethylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.