missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Chloro-N-methylacetanilide, 97%

$36.32 - $578.77

Chemical Identifiers

CAS 74585-34-5
Molecular Formula C9H10ClNO
Formula Weight 183.64
MDL Number MFCD01313707
Synonym N-(2-Chlorophenyl)-N-methylacetamide
Chemical Name or Material 2'-Chloro-N-methylacetanilide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2528203 SDS Alfa Aesar™
B2528203
1g Each for $36.32
AAB2528206 SDS Alfa Aesar™
B2528206
5g Each for $140.39
AAB2528214 SDS Alfa Aesar™
B2528214
25g Each for $578.77
Specifications Description & Specifications

Chemical Identifiers

74585-34-5
183.64
N-(2-Chlorophenyl)-N-methylacetamide
C9H10ClNO
MFCD01313707
2'-Chloro-N-methylacetanilide

Specifications

1g
C9H10ClNO
MFCD01313707
2'-Chloro-N-methylacetanilide
74585-34-5
183.64
N-(2-Chlorophenyl)-N-methylacetamide
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning