missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-N-methylacetamide 98.0+%, TCI America™
Supplier: TCI America C301425G
Specifications
| 2-Chloro-N-methylacetamide | |
| 44°C | |
| 116°C | |
| MFCD00018913 | |
| n-methyl-2-chloroacetamide, acetamide, 2-chloro-n-methyl, n-methylchloroacetamide, usaf do-35, 2-chloro-n-methyl-acetamide, alpha-chloro-n-methylacetamide, n-methyl 2-chloroacetamide, .alpha.-chloro-n-methylacetamide, n-methylchloracetamide, n-methyl chloroacetamide | |
| CNC(=O)CCl | |
| 107.537 | |
| 107.54 | |
| Crystalline Lumps |
| 96-30-0 | |
| White | |
| C3H6ClNO | |
| 25 g | |
| HOZLOOPIXHWKCI-UHFFFAOYSA-N | |
| 2-chloro-N-methylacetamide | |
| 66773 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 96-30-0 | |
| 107.537 | |
| HOZLOOPIXHWKCI-UHFFFAOYSA-N | |
| 66773 | |
| CNC(=O)CCl |
| C3H6ClNO | |
| MFCD00018913 | |
| n-methyl-2-chloroacetamide, acetamide, 2-chloro-n-methyl, n-methylchloroacetamide, usaf do-35, 2-chloro-n-methyl-acetamide, alpha-chloro-n-methylacetamide, n-methyl 2-chloroacetamide, .alpha.-chloro-n-methylacetamide, n-methylchloracetamide, n-methyl chloroacetamide | |
| 2-chloro-N-methylacetamide |
Safety and Handling
RTECSNumber : AB5775000
TSCA : No