Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-N-methoxy-N-methylacetamide, 98%

$100.97 - $321.72

Chemical Identifiers

Chemical Name or Material 2-Chloro-N-methoxy-N-methylacetamide
CAS 67442-07-3
Molecular Formula C4H8ClNO2
Formula Weight 137.57
MDL Number MFCD00134232
Beilstein 1924015
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1704806

 
alfa aesar™
L1704806
5g Each for $100.97

AAL1704814

 
alfa aesar™
L1704814
25g Each for $321.72
Description & Specifications

Specifications

Formula Weight 137.57
MDL Number MFCD00134232
CAS 67442-07-3
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 39° to 41°C
Quantity 5g
Assay 98%
Molecular Formula C4H8ClNO2
Beilstein 1924015
Health Hazard 1 Danger
Recommended Storage 2° to 8°C
Flash Point 104°C (219°F)
Boiling Point 94° to 95°C (9mmHg)
Chemical Name or Material 2-Chloro-N-methoxy-N-methylacetamide

Applications
2-Chloro-N-methoxy-N-methylacetamide is used in the preparation of 2-heptyl-3-hydroxy-4(1H)-quinolone (Pseudomonas quinolone signal or PQS) and structurally related 2-alkyl-4-quinolones having biological activity, 2-(benzo[d]thiazol-2-ylsulfonyl)-N-methoxy-N-methylacetamide and α-chloro-ketone, starting reagent for the one-pot synthesis of 2-heptyl-3-hydroxyl-4(1H)-quinolone (PQS), signaling molecule in the quorum sensing of Pseudomonas aeruginosa.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2-8°C. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C