Learn More
2-Chloro-N-methoxy-N-methylacetamide, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 67442-07-3 |
---|---|
Molecular Formula | C4H8ClNO2 |
Molecular Weight (g/mol) | 137.56 |
MDL Number | MFCD00134232 |
InChI Key | SCOJKGRNQDKFRP-UHFFFAOYSA-N |
Synonym | 2-chloro-n-methoxy-n-methyl acetamide, acetamide,2-chloro-n-methoxy-n-methyl, n-methoxy-n-methylchloroacetamide, acetamide, 2-chloro-n-methoxy-n-methyl, pubchem18657, acmc-209nz0, n-methyl-n-methoxy chloroacetamide, n-chloroacetyl-n-methoxymethylamine, n-methyl-n-methoxy-2-chloroacetamide, 2-chloro-n-methoxy-n-methyl-acetamide |
PubChem CID | 2734716 |
IUPAC Name | 2-chloro-N-methoxy-N-methylacetamide |
SMILES | CN(C(=O)CCl)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC369890050
|
Thermo Scientific Chemicals
369890050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
67442-07-3 | |
137.56 | |
SCOJKGRNQDKFRP-UHFFFAOYSA-N | |
2734716 | |
CN(C(=O)CCl)OC |
C4H8ClNO2 | |
MFCD00134232 | |
2-chloro-n-methoxy-n-methyl acetamide, acetamide,2-chloro-n-methoxy-n-methyl, n-methoxy-n-methylchloroacetamide, acetamide, 2-chloro-n-methoxy-n-methyl, pubchem18657, acmc-209nz0, n-methyl-n-methoxy chloroacetamide, n-chloroacetyl-n-methoxymethylamine, n-methyl-n-methoxy-2-chloroacetamide, 2-chloro-n-methoxy-n-methyl-acetamide | |
2-chloro-N-methoxy-N-methylacetamide |
Specifications
67442-07-3 | |
100.0 | |
104°C | |
Authentic | |
ClCH2CON(OCH3)CH3 | |
Glass bottle | |
2-chloro-n-methoxy-n-methyl acetamide, acetamide,2-chloro-n-methoxy-n-methyl, n-methoxy-n-methylchloroacetamide, acetamide, 2-chloro-n-methoxy-n-methyl, pubchem18657, acmc-209nz0, n-methyl-n-methoxy chloroacetamide, n-chloroacetyl-n-methoxymethylamine, n-methyl-n-methoxy-2-chloroacetamide, 2-chloro-n-methoxy-n-methyl-acetamide | |
CN(C(=O)CCl)OC | |
137.56 | |
137.56 | |
97% |
96.0 | |
94.0°C to 95.0°C | |
96% min. (GC) | |
C4H8ClNO2 | |
MFCD00134232 | |
5g | |
SCOJKGRNQDKFRP-UHFFFAOYSA-N | |
2-chloro-N-methoxy-N-methylacetamide | |
2734716 | |
Crystals | |
2-Chloro-N-methoxy-N-methylacetamide |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C
RUO â Research Use Only